Methyl-6-methylpiperidine-3-carboxylate
Catalog # APOSOR302409-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:157.21 g/mol
Storage Temperature:Ambient
CAS Number:908245-03-4
Formula:C₈H₁₅NO₂
MDL Number:MFCD13178893