

2-Methylbutyl isothiocyanate
Catalog # APOSOR30206-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:129.23 g/mol
Boiling Pt:84…86 °C/15mmHg
Storage Temperature:Ambient
UN:2927
ADR:6.1+8,II
:GHS05
:GHS06
CAS Number:4404-51-7
Formula:C₆H₁₁NS
MDL Number:MFCD00060414