
Methyl-3-bromothiophene-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26137-08-6
:
Formula:C₆H₅BrO₂S
MDL Number:MFCD00173839
Boiling Pt:112 °C/4mmHg
Melting Pt:50…51 °C
:221.07 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together