4'-Nitroacetophenone 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
- CAS Number:100-19-6
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007355
- Molecular Weight:165.15 g/mol
- Boiling Pt:201…202 °C
- Melting Pt:75…78 °C
- Flash Pt:149 °C
- Storage Temperature:Ambient
- EINECS:202-827-4
3 Options Available Below
Product Details & Documents
Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
- CAS Number:100-19-6
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007355
- Molecular Weight:165.15 g/mol
- Boiling Pt:201…202 °C
- Melting Pt:75…78 °C
- Flash Pt:149 °C
- Storage Temperature:Ambient
- EINECS:202-827-4



