Order Entry
Poland
ContactUsLinkComponent
(±)-1-Phenylethyl isothiocyanate
(±)-1-Phenylethyl isothiocyanate
Catalog # APOSOR28863-5G
Supplier:  Apollo Scientific
CAS Number:  
(±)-1-Phenylethyl isothiocyanate
Catalog # APOSOR28863-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :163.24 g/mol
Storage Temperature:Ambient
CAS Number:4478-92-6
 :32393-32-1
Formula:C₆H₅CH(NCS)CH₃
MDL Number:MFCD00022061