Order Entry
Poland
ContactUsLinkComponent
2,4-Xylyl isothiocyanate
Corrosive
Toxic
Health
2,4-Xylyl isothiocyanate
Catalog # APOSOR28861-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Toxic
Health
2,4-Xylyl isothiocyanate
Catalog # APOSOR28861-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :163.24 g/mol
Boiling Pt:122…130 °C/8mmHg
Melting Pt:25…29 °C
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,III
 :GHS05
 :GHS06
 :GHS08
CAS Number:39842-01-8
Formula:C₉H₉NS
MDL Number:MFCD00022054