Ethyl propiolate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:623-47-2
:
Formula:C₅H₆O₂
MDL Number:MFCD00009184
Boiling Pt:118…120 °C
Melting Pt:9 °C
Density:0.963 g/cm³ (25 °C)
Flash Pt:23 °C
Storage Temperature:Refrigerator
EINECS:210-795-8
ADR:3,III
:98.1 g/mol
UN:3272
:GHS02
:GHS07
Frequently Bought Together