Order Entry
Poland
ContactUsLinkComponent
2,6-Dibromo-p-toluidine
Irritant
2,6-Dibromo-p-toluidine
Catalog # APOSOR27443-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dibromo-p-toluidine
Catalog # APOSOR27443-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:72…76 °C
 :264.95 g/mol
EINECS:230-182-9
 :GHS07
Storage Temperature:Ambient
CAS Number:6968-24-7
Formula:C₇H₇Br₂N
MDL Number:MFCD00007641