Order Entry
Poland
Orders LinkContactUsLinkComponent
2,2-Dimethyloctane

2,2-Dimethyloctane

Bottle, 1 g
Catalog # APOSOR2370-1G
Flammable
Corrosive
Supplier:   Apollo Scientific
2,2-Dimethyloctane
Catalog # APOSOR2370-1G
Supplier:  Apollo Scientific
CAS Number:  15869-87-1
2,2-Dimethyloctane

Bottle, 1 g

CAS Number:  15869-87-1
Price
764 zł
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:15869-87-1
  • Formula:C₁₀H₂₂
  • Molecular Weight:142.28 g/mol
  • Boiling Pt:155 °C
  • Density:0.724 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:3295
  • ADR:3,III
  •  :Density: 0.7208 g/cm³ (25 °C)
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.