
1-(2,4-Dihydroxy-5-propylphenyl)ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:63411-87-0
:
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00100627
:194.23 g/mol
Melting Pt:99…101 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together