Order Entry
Poland
ContactUsLinkComponent
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 95+%
Irritant
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 95+%
Catalog # APOSOR17061-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 95+%
Catalog # APOSOR17061-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.58 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:84955-31-7
Formula:C₆H₅ClN₄
MDL Number:MFCD07369229