Order Entry
Poland
ContactUsLinkComponent
1-Boc-3-hydroxymethyl-5-methoxyindole 98%
Irritant
1-Boc-3-hydroxymethyl-5-methoxyindole 98%
Catalog # APOSOR1699-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Boc-3-hydroxymethyl-5-methoxyindole 98%
Catalog # APOSOR1699-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Boiling Pt:430.7 °C/760mmHg
Melting Pt:137…139 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:600136-09-2
Formula:C₁₅H₁₉NO₄