Order Entry
Poland
ContactUsLinkComponent
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
Irritant
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
Catalog # APOSOR14377-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
Catalog # APOSOR14377-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :156.19 g/mol
Melting Pt:78…81 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:258281-02-6
Formula:C₇H₁₂N₂O₂
MDL Number:MFCD03617955