Złóż zamówienie
Poland
Orders LinkContactUsLinkComponent
 

2,2-Dichloroacetamid

Irritant
New Product
Supplier:   Apollo Scientific
2,2-Dichloroacetamid
Supplier:  Apollo Scientific
New Product
Certificates
2,2-Dichloroacetamid

5 Options Available Below

Product Details & Documents
  • CAS Number:683-72-7
  • Formula:C₂H₃Cl₂NO
  • MDL Number:MFCD00008015
  • Molecular Weight:127.96 g/mol
  • Boiling Pt:233…234 °C
  • Melting Pt:96…101 °C
  • Storage Temperature:Temperatura otoczenia
  • EINECS:211-674-2
Product Family Options

Product Information

  • Rodzaj opakowania
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 11 080 zł
    11 080 zł
    1 * 2,5 kg
       11 080 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 149 zł
    149 zł
    1 * 25 g
       149 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 107 zł
    107 zł
    1 * 5 g
       107 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 580 zł
    580 zł
    1 * 100 g
       580 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 2 270 zł
    2 270 zł
    1 * 500 g
       2 270 zł

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.