
N-Methyl-4-boronobenzenesulphonamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:226396-31-2
:
Formula:C₇H₁₀BNO₄S
MDL Number:MFCD06659864
:215.03 g/mol
Melting Pt:136…140 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together