
3-Methoxy-4-methylbenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3556-60-3
:
Formula:C₉H₉NO
MDL Number:MFCD03428590
:147.1739 g/mol
Melting Pt:54 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
Frequently Bought Together