
4-(1-Pyrrolidinyl)piperidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5004-07-9
:
Formula:C₉H₁₈N₂
MDL Number:MFCD00038011
:154.2526 g/mol
Melting Pt:52…58 °C
EINECS:225-676-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together