
2-Isopropyl-6-methylphenyl isothiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306935-86-4
:
Formula:C₁₁H₁₃NO
MDL Number:MFCD00060370
:175.23 g/mol
Boiling Pt:92 °C/3mmHg
Density:1.02 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together