

3-Picolyl chloride hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6959-48-4
:
Formula:C₆H₇Cl₂N
MDL Number:MFCD00012818
:164.03 g/mol
Melting Pt:137…143 °C
Storage Temperature:Ambient
EINECS:230-150-4
UN:3261
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together