Order Entry
Poland
ContactUsLinkComponent
Aluminium oxide 99.999%
Aluminium oxide 99.999%
Catalog # APOSIN1039-100G
Supplier:  Apollo Scientific
CAS Number:  
Aluminium oxide 99.999%
Catalog # APOSIN1039-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :101.96 g/mol
Boiling Pt:2980 °C
Melting Pt:2072 °C
Density:3.97 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:1344-28-1
 :11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424