D-(-)-Tartaric acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:147-71-7
:
Formula:C₄H₆O₆
MDL Number:MFCD00004238
:150.09 g/mol
Melting Pt:166…174 °C
Density:1.76 g/cm³ (25 °C)
Flash Pt:210 °C
EINECS:205-695-6
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together