Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Midostaurin

Irritant
Supplier:   Apollo Scientific
Midostaurin
Supplier:  Apollo Scientific
Certificates

6 Options Available Below

Product Details & Documents

Semisynthetic derivate of Staurosporine. Inhibitor of PKC and cAMPK.

Synonyms:  PKC412, N-((5S,6R,7R,9R)-6-Methoxy-5-methyl-14-oxo-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-7-yl)-N-methylbenzamide
  • CAS Number:120685-11-2
  • Formula:C₃₅H₃₀N₄O₄
  • MDL Number:MFCD12828879
  • Molecular Weight:570.64 g/mol
  • Melting Pt:235…260 °C
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 665 zł
    665 zł
    1 * 5 mg
       665 zł

  • Bottle
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 180 zł
    1 180 zł
    1 * 10 mg
       1 180 zł

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 2 580 zł
    2 580 zł
    1 * 25 mg
       2 580 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 4 630 zł
    4 630 zł
    1 * 50 mg
       4 630 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 8 320 zł
    8 320 zł
    1 * 100 mg
       8 320 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 18 170 zł
    18 170 zł
    1 * 250 mg
       18 170 zł

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.