Paxilline (from Penicillium paxilli)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:57186-25-1
:
Formula:C₂₇H₃₃NO₄
MDL Number:MFCD00083464
Molecular Weight:435.564 g/mol
UN:2811
ADR:6.1 II
:GHS05
:GHS06
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together