
Fosfomycin disodium salt
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26016-99-9
:
Formula:C₃H₅Na₂O₄P
MDL Number:MFCD00056853
:182.02 g/mol
EINECS:247-409-2
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together