
Fluorescein isothiocyanate dextran
:60842-46-8
:
:MFCD00131092
:GHS07
:Refrigerator
Frequently Bought Together
2-Propanol ≥70% (v/v), SAFC®
: MERCK PRODUCTION CHEMICALS
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
: PanReac AppliChem
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
1-Chloro-2,4-dinitrobenzene 99%
: Thermo Fisher Scientific
Dextran, M.W. 150,000 TRITC-labeled
: AAT BIOQUEST
N,N-Dimethylformamide ≥99.8% ACS
: Thermo Fisher Scientific