Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Flavanomarein
Flavanomarein for HPLC
Flavanomarein for HPLC
Flavanomarein for HPLC
APOSBIDF1015-10MG
:
Apollo Scientific
:
Flavanomarein for HPLC
APOSBIDF1015-10MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
10 mg
Seq:
2275
:
:
450.39 g/mol
:
243…246 °C
:
209-411-1
:
GHS07
:
Ambient
:
577-38-8
:
C₂₁H₂₂O₁₁
:
MFCD00136058