Benzyl-2,3-O-isopropylidene-α-D-mannofuranoside 98% min
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20689-03-6
:
Formula:C₁₆H₂₂O₆
MDL Number:MFCD02683277
:310.346 g/mol
Storage Temperature:Ambient
Frequently Bought Together