

Sodium dodecyl sulphate (SDS) ≥99%, Ultrapure
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Melting Pt:204…207 °C
Density:0.37 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:205-788-1
UN:2926
ADR:4.1+6.1,II
:GHS02
:GHS06
Frequently Bought Together