Rabbit recombinant OX40 (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rabbit
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Gene ID:O02764
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- Protein/peptide name:OX40
- Purity:>85%
- Molecular weight:47,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Frequently Bought Together