Mouse recombinant CD155 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Mouse TIGIT, His Tag (Cat. No. ACRBTIT-M52E6) at 10 μg/ml (100 μl/well) can bind Mouse CD155, Mouse IgG2a Fc Tag (Cat. No. ACRBCD5-M5254) with a linear range of 0,039 - 1,25 μg/ml (QC tested)
- Gene ID:Q8K094-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
- Protein/peptide name:CD155
- Purity:>95%
- Molecular weight:61,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Specifications
Frequently Bought Together