Human recombinant SLAMF7 (from HEK293)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Humanized Anti-Human CS1 MAb at 1 μg/ml (100 µl/well) can bind Biotinylated Human SLAMF7, Avitag,His Tag with a linear range of 0,6-5 ng/ml (Routinely tested).; 2e5 of Anti-SLAMF7 CAR-293 cells were stained with 100 µl of 0,1 μg/ml of Biotinylated Human SLAMF7, Avitag, His Tag and negative control protein respectively, washed and then followed by PE-SA and analyzed with FACS (QC tested).
- Gene ID:Q9NQ25-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:FOAP-12|CRACC|19A|CS1|CD319|SLAMF7
- Protein/peptide name:SLAMF7
- Purity:>95%
- Molecular weight:25,6 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together