Order Entry
Poland
ContactUsLinkComponent
Mouse recombinant Fc gamma RI/ CD64 (from HEK293 cells)
Mouse recombinant Fc gamma RI/ CD64 (from HEK293 cells)
Catalog # ACRBCD4-M5227-50UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Mouse recombinant Fc gamma RI/ CD64 (from HEK293 cells)
Catalog # ACRBCD4-M5227-50UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag (BLI verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Captured Mouse CD64, His Tag (ACRBCD4-M5227) on NTA-Series S sensor chip can bind Herceptin® (Trastuzumab) with an affinity constant of 63,8 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Herceptin on Protein A Biosensor, can bind Mouse CD64, His Tag (ACRBCD4-M5227) with an affinity constant of 31 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P26151-1
  • Protein synonyms:
    FCGR1|FCG1|FCRI|FCGR1A|IGFR1|CD64|CD64A
  • Protein/peptide name:
    Fc gamma RI/ CD64
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    32,3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

Specifications

About this item

Mouse Fc gamma RI / CD64 Protein, His Tag (BLI verified).