Order Entry
Poland
ContactUsLinkComponent
 
Human recombinant MAG/ Siglec-4a (from HEK293 cells)
 
Human recombinant MAG/ Siglec-4a (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    P20916-1
  • Protein synonyms:
    GMA|MAG|Siglec-4a|S-MAG
  • Protein/peptide name:
    MAG/ Siglec-4a
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    81,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

 

Frequently Bought Together