Human recombinant B7-H5 (from HEK293)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Loaded Biotinylated Human B7-H5, Avitag,His Tag (Cat. No. B75-H82E1) on SA Biosensor, can bind Human VSIG3, Fc Tag (Cat. No. VS3-H5258) with an affinity constant of 0,312 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested)
- Gene ID:Q9H7M9-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:B7-H5|SISP1|Gi24|VISTA
- Protein/peptide name:B7-H5
- Purity:>95%
- Molecular weight:20,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Frequently Bought Together