Human recombinant OX40 (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation,; Measured by its binding ability in a functional ELISA, Immobilised Human OX40 Ligand, His Tag (Cat, No, OXL-H52Q8) at 2 μg/ml (100 μl/well) can bind Human OX40, Mouse IgG2a Fc Tag, low endotoxin (Cat, No, OX0-H5252) with a linear range of 0,78-25 ng/ml (QC tested),; FACS assay shows that recombinant Human OX40, Mouse IgG2a Fc Tag, low endotoxin (Cat, No, OX0-H5252) can bind to 293T cell overexpressing human OX40L, The concentration of OX40 is 0,01 μg/ml (Routinely tested).
- Gene ID:P43489-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TXGP1L|ACT35|OX40|TNFRSF4|OX40L receptor|CD134
- Protein/peptide name:OX40
- Purity:>95%
- Molecular weight:47,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together