Order Entry
Poland
ContactUsLinkComponent
2-[4-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxaldehyde
Irritant
2-[4-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxaldehyde
Catalog # APOSPC9656-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-[4-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxaldehyde
Catalog # APOSPC9656-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.232 g/mol
Melting Pt:103…105 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:174006-70-3
Formula:C₁₁H₆F₃NOS
MDL Number:MFCD07772870