Methyl 1-[2,6-dinitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylate 97%
Catalog # APOSPC9406-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:377.273 g/mol
Melting Pt:100…102 °C
Storage Temperature:Ambient
CAS Number:303144-40-3
Formula:C₁₄H₁₄F₃N₃O₆
MDL Number:MFCD00664468