
2,2-Bis(4-methacryloxyphenyl)-1,1,1,3,3,3-hexafluoropropane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:108050-42-6
:
Formula:C₂₃H₁₈F₆O₄
MDL Number:MFCD00080522
:472.377 g/mol
Melting Pt:116 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together