Order Entry
Poland
ContactUsLinkComponent
2-Chloro-4-(difluoromethyl)pyrimidine ≥98%
2-Chloro-4-(difluoromethyl)pyrimidine ≥98%
Catalog # APOSPC907312-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-(difluoromethyl)pyrimidine ≥98%
Catalog # APOSPC907312-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :164.541 g/mol
Storage Temperature:Ambient
CAS Number:1261629-31-5
Formula:C₅H₃ClF₂N₂
MDL Number:MFCD18415621