Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(5-Chloro-3-fluoropyridin-2-yl)methanol
(5-Chloro-3-fluoropyridin-2-yl)methanol 98%
(5-Chloro-3-fluoropyridin-2-yl)methanol 98%
(5-Chloro-3-fluoropyridin-2-yl)methanol 98%
APOSPC903907-250MG
:
Apollo Scientific
:
(5-Chloro-3-fluoropyridin-2-yl)methanol 98%
APOSPC903907-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
161.561 g/mol
:
GHS07
:
Refrigerator
:
214055-12-6
:
C₆H₅ClFNO
:
MFCD16607027