N,N-Dimethyl-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Catalog # APOSPC8876-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:346.325 g/mol
Storage Temperature:Ambient
CAS Number:1227954-79-1
Formula:C₁₄H₁₃F₃N₂O₃S
MDL Number:MFCD11109526