Order Entry
Poland
ContactUsLinkComponent
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Catalog # APOSPC8874-1G
Supplier:  Apollo Scientific
CAS Number:  
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Catalog # APOSPC8874-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :346.325 g/mol
Storage Temperature:Ambient
CAS Number:1257535-24-2
Formula:C₁₄H₁₃F₃N₂O₃S
MDL Number:MFCD11109524