Order Entry
Poland
ContactUsLinkComponent
 
Irritant
N-[4-(Trifluoromethyl)pyrimidin-2-yl]ethane-1,2-diamine
Catalog # APOSPC8813-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-[4-(Trifluoromethyl)pyrimidin-2-yl]ethane-1,2-diamine
Catalog # APOSPC8813-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.168 g/mol
Boiling Pt:104…106 °C (0.75 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:215655-29-1
Formula:C₇H₉F₃N₄
MDL Number:MFCD00816843