
1H,1H-Heptadecafluoro(2,5-dimethyl-3,6-dioxanonan-1-ol)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14548-74-4
:
Formula:C₉H₃F₁₇O₃
MDL Number:MFCD00339457
Boiling Pt:155…156 °C
Density:1.8 g/cm³ (25 °C)
:482.091 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together