
1H,1H,2H,2H-Perfluoro-9-methyldecan-1-ol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:31200-98-3
:
Formula:C₁₁H₅F₁₉O
MDL Number:MFCD00198022
:514.126 g/mol
Boiling Pt:100 °C (7 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together