
1H,1H,1H,2H,2H-Perfluorooctane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:80793-17-5
:
Formula:C₈H₅F₁₃
MDL Number:MFCD00236705
:348.105 g/mol
Boiling Pt:114…116 °C
Density:1.566 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together