
4-(Difluoromethyl)aniline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1980063-54-4
:
Formula:C₇H₇F₂N·ClH
MDL Number:MFCD28145075
:179.595 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together