Order Entry
Poland
ContactUsLinkComponent
1-Bromo-5-chloro-1,1,3,3,5,5-hexafluoropentane
Irritant
1-Bromo-5-chloro-1,1,3,3,5,5-hexafluoropentane
Catalog # APOSPC56223-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Bromo-5-chloro-1,1,3,3,5,5-hexafluoropentane
Catalog # APOSPC56223-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :293.433 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1980045-73-5
Formula:C₅H₄BrClF₆
MDL Number:MFCD28901029