
5-(Difluoromethyl)pyridine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:859538-41-3
:
Formula:C₇H₅F₂NO₂
MDL Number:MFCD18256209
:173.117 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together