Order Entry
Poland
ContactUsLinkComponent
Methyl 5-chloro-2-fluoro-4-hydroxybenzoate ≥98%
Methyl 5-chloro-2-fluoro-4-hydroxybenzoate ≥98%
Catalog # APOSPC540017-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5-chloro-2-fluoro-4-hydroxybenzoate ≥98%
Catalog # APOSPC540017-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :204.583 g/mol
Storage Temperature:Refrigerator
CAS Number:245743-64-0
Formula:C₈H₆ClFO₃
MDL Number:MFCD22371002